Learn More
N-Desmethyltramadol Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-Desmethyltramadol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-Desmethyl-cis-tramadol Hydrochloride |
| CAS | 1018989-94-0 |
| Analyte or Component Names | N-Desmethyl-cis-tramadol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-Desmethyltramadol Hydrochloride, Cyclohexanol, 1-(3-methoxyphenyl)-2-[(methylamino)methyl]-, hydrochloride (1:1), (1R,2R)-rel-, N-Demethyltramadol hydrochloride |
| InChI Formula | InChI=1S/C15H23NO2.ClH/c1-16-11-13-6-3-4-9-15(13,17)12-7-5-8-14(10-12)18-2;/h5,7-8,10,13,16-17H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1 |
| SMILES | Cl.CNC[C@H]1CCCC[C@]1(O)c2cccc(OC)c2 |
| IUPAC Name | (1R,2R)-1-(3-methoxyphenyl)-2-(methylaminomethyl)cyclohexan-1-ol;hydrochloride |
| Show More |