missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Clotiazepam, TRC

Product Code. 30138194
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138194 100 mg 100mg
17903265 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138194 Supplier Toronto Research Chemicals Supplier No. TRCD291535100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33671-37-3
Molecular Formula C15 H13 Cl N2 O S
Molecular Weight (g/mol) 304.79
Synonym 5-(2-Chlorophenyl)-7-ethyl-1,3-dihydro-2H-thieno[2,3-e][1,4]diazepin-2-one, 5-(o-Chlorophenyl)-7-ethyl-1,3-dihydro-2H-thieno[2,3-e]-1,4-diazepin-2-one
IUPAC Name 5-(2-chlorophenyl)-7-ethyl-1,3-dihydrothieno[2,3-e][1,4]diazepin-2-one
SMILES CCc1cc2C(=NCC(=O)Nc2s1)c3ccccc3Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H13ClN2OS/c1-2-9-7-11-14(10-5-3-4-6-12(10)16)17-8-13(19)18-15(11)20-9/h3-7H,2,8H2,1H3,(H,18,19)
Formula Weight 304.0437
Chemical Name or Material N-Desmethyl Clotiazepam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.