CAS RN 33671-37-3
CAS RN 33671-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Desmethyl Clotiazepam, TRC
CAS: 33671-37-3 Molecular Formula: C15 H13 Cl N2 O S Molecular Weight (g/mol): 304.79 Synonym: 5-(2-Chlorophenyl)-7-ethyl-1,3-dihydro-2H-thieno[2,3-e][1,4]diazepin-2-one,5-(o-Chlorophenyl)-7-ethyl-1,3-dihydro-2H-thieno[2,3-e]-1,4-diazepin-2-one IUPAC Name: 5-(2-chlorophenyl)-7-ethyl-1,3-dihydrothieno[2,3-e][1,4]diazepin-2-one SMILES: CCc1cc2C(=NCC(=O)Nc2s1)c3ccccc3Cl