Learn More
N-Deacetyl-N-formyl Colchicine, TRC
Product Code. 30011435
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011435 50 mg 50mg
30007883 25 mg 25mg
30003282 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011435 Supplier Toronto Research Chemicals Supplier No. TRCD19894050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7411-12-3
Molecular Formula C21 H23 N O6
Molecular Weight (g/mol) 385.41
Synonym N-[(7S,12aRa)-1,2,3,10-tetramethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]formamide, N-[(7S)-5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl]-formamide, Colchicine, N-deacetyl-N-formyl- (7CI,8CI), Colchicine, deacetyl-N-formyl- (6CI), Formamide, N-(5,6,7,9-tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)-, (S)-, Benzo[a]heptalene, formamide deriv., Gloriosine, N-Deacetyl-N-formylcolchicine, N-Formyl-N-deacetylcolchicine, N-Formyl-N-desacetylcolchicine, N-Formyldeacetylcolchicine, NSC 403142, Colchicine Imp. A (EP)
IUPAC Name N-[(7S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]formamide
SMILES COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C21H23NO6/c1-25-17-8-6-13-14(10-16(17)24)15(22-11-23)7-5-12-9-18(26-2)20(27-3)21(28-4)19(12)13/h6,8-11,15H,5,7H2,1-4H3,(H,22,23)/t15-/m0/s1
Formula Weight 385.1525
Percent Purity >95%
Chemical Name or Material N-Deacetyl-N-formyl Colchicine
Grade HPLC