missing translation for 'onlineSavingsMsg'
Learn More

N-(Cyanomethyl)pyridinium chloride, 97%

Produktkod. p-7021830
Change view
Klicka för att se tillgängliga alternativ
Quantity:
10 g
50 g
250 g
Förpackningsstorlek:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11476877 10 g 10g
11486877 50 g 50g
11496877 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11476877 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A14327.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 17281-59-3 | C7H7ClN2 | 154.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 17281-59-3
Molecular Formula C7H7ClN2
Molecular Weight (g/mol) 154.60
MDL Number MFCD00031933
InChI Key HEJOROWXIWLCMS-UHFFFAOYSA-M
Synonym 1-cyanomethyl pyridinium chloride, n-cyanomethyl pyridinium chloride, 1-cyanomethyl pyridin-1-ium chloride, acmc-1chuo, cyanomethylpyridinium chloride, n-cyanomethyl pyridiniumchloride, 2-pyridin-1-ium-1-ylacetonitrile chloride, 2-1-pyridin-1-iumyl acetonitrile chloride, 2-pyridin-1-ium-1-ylethanenitrile chloride
PubChem CID 2734116
IUPAC Name 2-pyridin-1-ium-1-ylacetonitrile;chloride
SMILES [Cl-].N#CC[N+]1=CC=CC=C1

Specifikationer

Melting Point 173°C to 175°C
Quantity 10 g
UN Number UN3439
Beilstein 3728392
Formula Weight 154.6
Percent Purity 97%
Chemical Name or Material N-(Cyanomethyl)pyridinium chloride

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.