Learn More
N-CBZ-Glycyl-glycyl-L-arginine 7-Amido-4-methylcoumarin Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-CBZ-Glycyl-glycyl-L-arginine 7-Amido-4-methylcoumarin Hydrochloride |
| CAS | 102601-58-1 |
| Molecular Formula | C28H33N7O7 . HCl |
| Quantity | 5 mg |
| Synonym | N-[(Phenylmethoxy)carbonyl]glycylglycyl-N-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-L-argininamide Monohydrochloride |
| InChI Formula | InChI=1S/C28H33N7O7.ClH/c1-17-12-25(38)42-22-13-19(9-10-20(17)22)34-26(39)21(8-5-11-31-27(29)30)35-24(37)15-32-23(36)14-33-28(40)41-16-18-6-3-2-4-7-18;/h2-4,6-7,9-10,12-13,21H,5,8,11,14-16H2,1H3,(H,32,36)(H,33,40)(H,34,39)(H,35,37)(H4,29,30,31);1H/t21-;/m0./s1 |
| SMILES | ClC1=CC(=O)C=C(O1)N2CCOCC2 |
| IUPAC Name | benzyl N-[2-[[2-[[(2S)-5-carbamimidamido-1-[(4-methyl-2-oxochromen-7-yl)amino]-1-oxopentan-2-yl]amino]-2-oxoethyl]amino]-2-oxoethyl]carbamate;hydrochloride |
| Molecular Weight (g/mol) | 579.6 |
| Formula Weight | 615.2208 |