Learn More
N-Carboxybenzyl Valbenazine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Carboxybenzyl Valbenazine |
| CAS | 1025504-76-0 |
| Molecular Formula | C32H44N2O6 |
| Quantity | 10 mg |
| Synonym | N-[(Phenylmethoxy)carbonyl]-L-Valine (2R,3R,11bR)-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-yl Ester |
| InChI Formula | InChI=1S/C32H44N2O6/c1-20(2)14-24-18-34-13-12-23-15-28(37-5)29(38-6)16-25(23)26(34)17-27(24)40-31(35)30(21(3)4)33-32(36)39-19-22-10-8-7-9-11-22/h7-11,15-16,20-21,24,26-27,30H,12-14,17-19H2,1-6H3,(H,33,36)/t24-,26-,27-,30+/m1/s1 |
| SMILES | COc1cc2CCN3C[C@@H](CC(C)C)[C@@H](C[C@@H]3c2cc1OC)OC(=O)[C@@H](NC(=O)OCc4ccccc4)C(C)C |
| IUPAC Name | [(2R,3R,11bR)-9,10-dimethoxy-3-(2-methylpropyl)-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizin-2-yl] (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate |
| Molecular Weight (g/mol) | 552.7 |
| Formula Weight | 552.3199 |