Learn More
n-Butyl benzoate, 98%
Product Code. 11418866
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
Product Code. Quantity unitSize
11418866 250 g 250g
11408866 1000 g 1000g
2 options
Product Code. 11418866
Brand: Thermo Scientific Alfa Aesar A17010.30
Request Bulk or Custom Format
CAS: 136-60-7 | C11H14O2 | 178.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 136-60-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00009439
InChI Key XSIFPSYPOVKYCO-UHFFFAOYSA-N
Synonym n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
PubChem CID 8698
IUPAC Name butyl benzoate
SMILES CCCCOC(=O)C1=CC=CC=C1
Specifications
Melting Point -22°C
Density 1.011
Boiling Point 250°C
Flash Point 106°C (222°F)
Refractive Index 1.498
Quantity 250 g
Beilstein 1867073
Merck Index 14,1552
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material n-Butyl benzoate