Learn More
N-Butyl-2-pyrrolidone, Dr. Ehrenstorfer
Product Code. 30316222
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316222 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30316222 Supplier Dr. Ehrenstorfer Supplier No. DRECA10931640
 Restricted Item
Estimated Shipment: 01-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mL
Certifications/Compliance ISO/IEC 17025