missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid, 98%

Product Code. p-7047593
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378836 1 g 1g
11388836 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378836 Supplier Thermo Scientific Alfa Aesar Supplier No. L19488.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51077-16-8 | C9H15NO4S | 233.282 g/mol

N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51077-16-8
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.282
MDL Number MFCD00077002
InChI Key FJWNZTPXVSWUKF-LURJTMIESA-N
Synonym boc-thz-oh, 4r-3-tert-butoxycarbonyl-1,3-thiazolane-4-carboxylic acid, n-boc-r-thiazolidine-4-carboxylic acid, 4r-3-tert-butoxycarbonyl-1,3-thiazolidine-4-carboxylic acid, r-3-tert-butoxycarbonyl thiazolidine-4-carboxylic acid, 4r-3-tert-butoxy carbonyl-1,3-thiazolidine-4-carboxylic acid, 3,4-thiazolidinedicarboxylicacid, 3-1,1-dimethylethyl ester, 4r, boc-thioproline, n-boc-thioproline, --boc-l-thioproline
PubChem CID 688405
IUPAC Name (4R)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CSCC1C(=O)O

Specifications

Melting Point ∽133°C (decomposition)
Quantity 1 g
Beilstein 6686084
Solubility Information Insoluble in water.
Optical Rotation −113° (c=1 in Methanol)
Formula Weight 233.28
Percent Purity 98%
Chemical Name or Material N-Boc-(R)-(-)-thiazolidine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.