missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-prolinal, 96%

Product Code. 11445870
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445870 1 g 1g
11455870 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445870 Supplier Thermo Scientific Alfa Aesar Supplier No. H26495.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 69610-41-9 | C10H17NO3 | 199.25 g/mol

Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.

Solubility
Slightly soluble water. Soluble in chloroform.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69610-41-9
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00274186,MFCD00801206
InChI Key YDBPZCVWPFMBDH-QMMMGPOBSA-N
Synonym n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
PubChem CID 2734674
IUPAC Name tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1C=O

Specifications

Density 1.06
Boiling Point 228°C
Flash Point >110°C (230°F)
Refractive Index 1.4589
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Slightly soluble water. Soluble in chloroform.
Optical Rotation −95° (c=1 in Chloroform)
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material N-Boc-L-prolinal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.