missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-isoleucine hemihydrate, 98+%

Product Code. 15487727
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487727 25 g 25g
15467727 1 g 1g
15477727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487727 Supplier Alfa Aesar™ Supplier No. H37050.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 204138-23-8
Molecular Formula C22H44N2O9
Molecular Weight (g/mol) 480.599
MDL Number MFCD00672705
InChI Key NYGJSARLXBZKTA-AQHOMMPKSA-N
Synonym boc-l-isoleucine hemihydrate, boc-ile-oh 1/2h2o, boc-isoleucine hemihydrate, boc-l-isoleucine 1/2h2o, n-boc-l-isoleucine hemihydrate, n-boc-l-isoleucine hemihydrate, 98+%, n-t-butyloxycarbonyl-l-isoleucine hemihydrate, n-tert-butoxycarbonyl-l-isoleucine hemihydrate, n-alpha-t-butyloxycarbonyl-l-isoleucine semihydrate, l-isoleucine, n-1,1-dimethylethoxy carbonyl-, hydrate 2:1
PubChem CID 45072434
IUPAC Name (2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
SMILES CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.O

Specifications

Melting Point 67°C to 70°C
Quantity 25 g
Formula Weight 240.07 (231.06 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-isoleucine hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.