Learn More
Alfa Aesar™ N-Boc-L-isoleucine hemihydrate, 98+%
Product Code. 15467727
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467727 1 g 1g
15477727 5 g 5g
15487727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15467727 Supplier Alfa Aesar™ Supplier No. H37050.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 204138-23-8
Molecular Formula C22H44N2O9
Molecular Weight (g/mol) 480.599
MDL Number MFCD00672705
InChI Key NYGJSARLXBZKTA-AQHOMMPKSA-N
Synonym boc-l-isoleucine hemihydrate, boc-ile-oh 1/2h2o, boc-isoleucine hemihydrate, boc-l-isoleucine 1/2h2o, n-boc-l-isoleucine hemihydrate, n-boc-l-isoleucine hemihydrate, 98+%, n-t-butyloxycarbonyl-l-isoleucine hemihydrate, n-tert-butoxycarbonyl-l-isoleucine hemihydrate, n-alpha-t-butyloxycarbonyl-l-isoleucine semihydrate, l-isoleucine, n-1,1-dimethylethoxy carbonyl-, hydrate 2:1
PubChem CID 45072434
IUPAC Name (2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
SMILES CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.O
Specifications
Melting Point 67°C to 70°C
Quantity 1 g
Formula Weight 240.07 (231.06 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-isoleucine hemihydrate