Learn More
N-Boc-L-glutamic Acid 5-Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-Boc-L-glutamic Acid 5-Methyl Ester |
| CAS | 45214-91-3 |
| Molecular Formula | C11H19NO6 |
| Quantity | 1 g |
| Synonym | N-tert-Butoxycarbonylglutamic Acid gamma-Methyl Ester, N-[(1,1-Dimethylethoxy)carbonyl]-L-glutamic Acid 5-Methyl Ester |
| InChI Formula | InChI=1S/C11H19NO6/c1-11(2,3)18-10(16)12-7(9(14)15)5-6-8(13)17-4/h7H,5-6H2,1-4H3,(H,12,16)(H,14,15)/t7-/m0/s1 |
| SMILES | COC(=O)CC[C@H](NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | (2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
| Molecular Weight (g/mol) | 261.27 |
| Formula Weight | 261.1212 |