Learn More
n-Benzyl-tert-butylamine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 3378-72-1 |
| Melting Point | -30°C to -25°C |
| Density | 0.90 g/cm3 at 20°C |
| Molecular Formula | C6H5CH2NHC(CH3)3 |
| MDL Number | MFCD00008798 |
| Quantity | 250 mL |
| Synonym | N-Benzyl-tert-butylamine, N-tert-Butylbenzylamine, N-Benzyl-tert-butylamine |
| Molecular Weight (g/mol) | 163.26 |
| Grade | Research |