missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-methoxybenzylamine, 97%

Product Code. p-7037065
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411948 250 mg 250mg
15401948 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15411948 Supplier Thermo Scientific Alfa Aesar Supplier No. H55633.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14429-02-8 | C15H17NO | 227.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14429-02-8
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.31
MDL Number MFCD01002920
InChI Key WZUBICZIDUTTNJ-UHFFFAOYSA-N
Synonym benzyl 4-methoxyphenyl methyl amine, benzyl-4-methoxy-benzyl-amine, n-benzyl-1-4-methoxyphenyl methanamine, benzyl-4-methoxybenzyl amine, n-benzyl-4-methoxybenzylamine, n-benzyl-n-4-methoxybenzyl amine, 4-methoxyphenyl methyl benzylamine, 4-methoxybenzylbenzylamine, cbmicro_025161, cbmicro_047998
PubChem CID 738564
IUPAC Name N-[(4-methoxyphenyl)methyl]-1-phenylmethanamine
SMILES COC1=CC=C(CNCC2=CC=CC=C2)C=C1

Specifications

Quantity 250 mg
UN Number UN3259
Formula Weight 227.31
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-methoxybenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.