missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl bromide, 90+%

Product Code. 11361547
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361547 100 g 100g
11351547 25 g 25g
11371547 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11361547 Supplier Thermo Scientific Alfa Aesar Supplier No. L02211.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 776-74-9 | C13H11Br | 247.135 g/mol

Benzhydryl bromide is a halogenated building block. Benzhydryl group was preferred to the commoner benzyl to protect 2-nitrophenol in the Bartoli (vinyl Grignard) synthesis of 7-hydroxyindole. Protection was by reaction with the phenol in the presence of potassium carbonate in acetone; deprotection of the indole was by hydrogenolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydryl bromide is a halogenated building block. Benzhydryl group was preferred to the commoner benzyl to protect 2-nitrophenol in the Bartoli (vinyl Grignard) synthesis of 7-hydroxyindole. Protection was by reaction with the phenol in the presence of potassium carbonate in acetone; deprotection of the indole was by hydrogenolysis.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Protect from humidity and water. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-74-9
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.135
MDL Number MFCD00000134
InChI Key OQROAIRCEOBYJA-UHFFFAOYSA-N
Synonym bromodiphenylmethane, benzhydryl bromide, diphenylmethyl bromide, diphenylbromomethane, alpha-bromodiphenylmethane, bromo phenyl methyl benzene, benzene, 1,1'-bromomethylene bis, methane, bromodiphenyl, bromomethylene dibenzene, bromodiphenyl methane
PubChem CID 236603
IUPAC Name [bromo(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Br

Specifications

Melting Point 36°C to 41°C
Boiling Point 183°C to 185°C (20 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 100 g
UN Number UN1770
Beilstein 638544
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 247.14
Percent Purity ≥90%
Chemical Name or Material Benzhydryl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.