Learn More
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%
Product Code. 15489725
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489725 5 g 5g
15499725 250 mg 250mg
15479725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15489725 Supplier Thermo Scientific Alfa Aesar Supplier No. H52309.06
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
CAS: 51549-39-4 | C23H31N5O4Si | 469.62 g/mol
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxyadenosine is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51549-39-4
Molecular Formula C23H31N5O4Si
Molecular Weight (g/mol) 469.62
MDL Number MFCD04972281
InChI Key BJZZQJFYKQPUGO-TWAUYLRJNA-N
IUPAC Name N-{9-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-9H-purin-6-yl}benzamide
SMILES CC(C)(C)[Si](C)(C)OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12
Specifications
Quantity 5 g
Formula Weight 469.62
Percent Purity ≥98%
Chemical Name or Material N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine