Learn More
N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid Sodium Salt, TRC
Product Code. 30313758
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313758 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30313758 Supplier Toronto Research Chemicals Supplier No. TRCA609250500MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid Sodium Salt
CAS 100900-07-0
Molecular Formula C12 H13 N2 O3 S . Na
Quantity 500 mg
InChI Formula InChI=1S/C12H14N2O3S.Na/c13-7-8-14-11-5-1-4-10-9(11)3-2-6-12(10)18(15,16)17;/h1-6,14H,7-8,13H2,(H,15,16,17);/q;+1/p-1
SMILES [Na+].NCCNc1cccc2c(cccc12)S(=O)(=O)[O-]
IUPAC Name sodium;5-(2-aminoethylamino)naphthalene-1-sulfonate
Molecular Weight (g/mol) 288.3
Formula Weight 288.0545