Learn More
N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | N-(Aminoethyl)-5-naphthylamine-1-sulfonic Acid Sodium Salt |
| CAS | 100900-07-0 |
| Molecular Formula | C12 H13 N2 O3 S . Na |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C12H14N2O3S.Na/c13-7-8-14-11-5-1-4-10-9(11)3-2-6-12(10)18(15,16)17;/h1-6,14H,7-8,13H2,(H,15,16,17);/q;+1/p-1 |
| SMILES | [Na+].NCCNc1cccc2c(cccc12)S(=O)(=O)[O-] |
| IUPAC Name | sodium;5-(2-aminoethylamino)naphthalene-1-sulfonate |
| Molecular Weight (g/mol) | 288.3 |
| Formula Weight | 288.0545 |