missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic acid, MP Biomedicals™

Product Code. 11487292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487292 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487292 Supplier MP Biomedicals Supplier No. 0215385191
  • ONLINE EXCLUSIVE PRICE
    1745.00 EUR
    1660.05 EUR / 1g
    Save 84.95 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N-Acetylneuraminic acid ≥98%, Off-White Powder

  • N-Acetylneuraminic acid (NANA, Neu5Ac) is a major component of glycoconjugates such as glycolipids, glycoproteins and proteoglycans (sialogylcoproteins) where it confers selective binding characteristics to the glycosylated component.
  • Neu5Ac is used to study its biochemistry, metabolism and uptake in vivo and in vitro. Neu5Ac is used in the development of nanocarriers.
  • Both sialic acid and neuraminic acid are loosely used to refer to conjugates of neuraminic acid. N-Acetylneuraminic acid is often found as the terminal sugar of cell surface glycoproteins.
  • Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion.
  • Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion.

Specifications

CAS 131-48-6
Melting Point 180°C to 190°C
Molecular Formula C11H19NO9
MDL Number MFCD00006620
Quantity 1 g
Beilstein 1716283
Synonym Sialic Acid, NANA, Type IV, 5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid
Solubility Information Soluble in water
InChI Key SQVRNKJHWKZAKO-UHFFFAOYNA-N
Molecular Weight (g/mol) 309.27
Physical Form Powder
Grade >98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.