missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylneuraminic Acid, >98%, MP Biomedicals™

Product Code. 11477292 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477292 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477292 Supplier MP Biomedicals Supplier No. 0215385125
  • ONLINE EXCLUSIVE PRICE
    95.90 EUR
    91.35 EUR / 25mg
    Save 4.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 131-48-6
Color White
Assay Percent Range >98%
Molecular Formula C11H19NO9
Quantity 25 mg
Synonym n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
InChI Key SQVRNKJHWKZAKO-LFIUDZTESA-N
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
IUPAC Name (4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
Molecular Weight (g/mol) 309.271
PubChem CID 126963458
Formula Weight 309.27
Physical Form Powder
Chemical Name or Material N-Acetylneuraminic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.