missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ N-Acetylgalactosamine-6-Sulfatase/GALNS Recombinant Protein Antigen
Shop All Bio Techne ProductsDescription
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GALNS. The N-Acetylgalactosamine-6-Sulfatase/GALNS Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 2588 |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | chondroitinase, chondroitinsulfatase, EC 3.1.6, EC 3.1.6.4, FLJ17434, FLJ42844, FLJ98217, galactosamine (N-acetyl)-6-sulfate sulfatase, Galactose-6-sulfate sulfatase, GALNAC6S, GalNAc6S sulfatase, GAS, MPS4A, N-acetylgalactosamine-6-sulfatase, N-acetylgalactosamine-6-sulfate sulfatase |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Symbol | GALNS |
| Molecular Weight (g/mol) | 28 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54401. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?