Learn More
Tocris Bioscience™ N-Acetylcysteine amide
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
N-Acetylcysteine amide is a glutathione (GSH) precursor and cell-permeable antioxidant. N-Acetylcysteine amide replenishes intracellular Glutathione (GSH). N-Acetylcysteine amide directly reduces intracellular Glutathione disulfide (GSSG) to GSH without glutathione peroxidase. N-Acetylcysteine amide has anti-inflammatory activity through regulation of activation of NF-κB and HIF-1α, as well as modulation of reactive oxygen species. N-Acetylcysteine amide improves neuronal mitochondrial bioenergetics, reduces tissue damage and enhances functional recovery following spinal cord injury in rats. N-Acetylcysteine amide also enhances behavioral recovery in rats following traumatic brain injury. Neuroprotective.
For more information about how N-Acetylcysteine amide may be used, see our protocol: 3D Culture of Lung Alveolar Cells
Specifications
Specifications
| Chemical Name or Material | (2R)-2-(Acetylamino)-3-mercaptopropanamide |
| CAS | 38520-57-9 |
| Quantity | 10 mg |
| Target | Additional Antioxidant Compounds |
| Molecular Formula | C5H10N2O2S |
| Formula Weight | 162.21 |