missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-arginine dihydrate, MP BIOMEDICALS

Product Code. 11318379 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318379 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11318379 Supplier MP Biomedicals Supplier No. 0210007805
  • ONLINE EXCLUSIVE PRICE
    80.50 EUR
    76.65 EUR / 5g
    Save 3.85 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Usage Statement:

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Specifications

CAS 210545-23-6
Color White
Molecular Formula C8H16N4O3 2H2O
Synonym 2-Acetamido-5-Guanidinovaleric Acid
Molecular Weight (g/mol) 216.241 g/mol
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.