missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-DL-tryptophan, ∽99%, MP Biomedicals™

Product Code. 11358459 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358459 1 kg 1kg
11338459 25 g 25g
11348459 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358459 Supplier MP Biomedicals Supplier No. 0210014291
  • ONLINE EXCLUSIVE PRICE
    1215.00 EUR
    1155.00 EUR / 1kg
    Save 60.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N-acetyl-DL-tryptophan, is used as stabilizer in the human blood-derived therapeutic products normal serum albumin and plasma protein fraction

Chemical Identifiers

CAS 87-32-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00005644
InChI Key DZTHIGRZJZPRDV-UHFFFAOYSA-N
Synonym n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
PubChem CID 2002
ChEBI CHEBI:70976
IUPAC Name 2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White
Assay Percent Range ∽99%
Quantity 1 kg
Formula Weight 246.3
Percent Purity ≥99%
Grade Research
Physical Form Powder
Chemical Name or Material N-acetyl-DL-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.