missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-d3-S-(3-hydroxypropyl-1-methyl)-L-cysteine Dicyclohexylammonium Salt, TRC

Product Code. 17908739
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908739 10 mg 10mg
17927199 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17908739 Supplier Toronto Research Chemicals Supplier No. TRCA17912710MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240483-48-0
Molecular Formula C21H37D3N2O4S
Molecular Weight (g/mol) 419.64
Synonym 3-Hydroxy-1-methylpropylmercapturic-N-acetyl-d3 Acid Dicyclohexylammonium Salt
SMILES OCCC(SC[C@@H](C([O-])=O)NC(C([2H])([2H])[2H])=O)C.C1([NH2+]C2CCCCC2)CCCCC1

Specifications

Quantity 10 mg
Chemical Name or Material N-Acetyl-d3-S-(3-hydroxypropyl-1-methyl)-L-cysteine Dicyclohexylammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.