missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-Glucosamine, MP Biomedicals™

Product Code. 11461585 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11461585

Brand: MP Biomedicals 0210006810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s). N-Acetyl-D-Glucosamine, MP Biomedicals is a derivitized glucose monomer found in polymers of bacterial cell walls, chitin, hyaluronic acids and various glycans.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7512-17-6
Melting Point 199°C (literature)
pH 7.20 (literature)
Molecular Formula C8H15NO6
MDL Number MFCD00061615
Quantity 10 g
Synonym 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
Solubility Information Soluble in water (50mg/mL).
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular Weight (g/mol) 221.21
Formula Weight 221.21
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.