missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-Glucosamine, Cell Culture Reagent, 100%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Color | White |
| Assay Percent Range | 1 |
| Quantity | 5 g |
| Formula Weight | 221.2 |
| Physical Form | Fine Crystalline Powder |
| Chemical Name or Material | N-Acetyl-D-Glucosamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction