Learn More
N,5-Dibenzyl-5-hydroxy-indole-3-glyoxylamide, TRC
Specifications
Specifications
| Chemical Name or Material | N,5-Dibenzyl-5-hydroxy-indole-3-glyoxylamide |
| CAS | 102754-41-6 |
| Molecular Formula | C24H20N2O3 |
| Quantity | 250 mg |
| Synonym | N-Benzyl-5-(benzyloxy)-indole-3-glyoxylamide, α-Oxo-5-(phenylmethoxy)-N-(phenylmethyl)-1H-Iindole-3-acetamide |
| InChI Formula | InChI=1S/C24H20N2O3/c27-23(24(28)26-14-17-7-3-1-4-8-17)21-15-25-22-12-11-19(13-20(21)22)29-16-18-9-5-2-6-10-18/h1-13,15,25H,14,16H2,(H,26,28) |
| SMILES | O=C(C(NCC1=CC=CC=C1)=O)C2=CNC3=CC=C(OCC4=CC=CC=C4)C=C32 |
| IUPAC Name | N-benzyl-2-(5-(benzyloxy)-1H-indol-3-yl)-2-oxoacetamide |
| Molecular Weight (g/mol) | 384.43 |
| Formula Weight | 384.15 |