Learn More
N-(4-Oxo-2-(1H-tetrazol-5-yl)-4H-chromen-8-yl)-4-(4-phenylbutoxy)benzamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 103177-37-3 |
| Molecular Formula | C27H23N5O4 |
| MDL Number | MFCD00864631 |
| Quantity | 250 mg |
| SMILES | O=C(NC1=C2C(C(C=C(C3=NN=NN3)O2)=O)=CC=C1)C4=CC=C(OCCCCC5=CC=CC=C5)C=C4 |
| Molecular Weight (g/mol) | 481.50 |
| Percent Purity | 98% |
| Chemical Name or Material | N-(4-Oxo-2-(1H-tetrazol-5-yl)-4H-chromen-8-yl)-4-(4-phenylbutoxy)benzamide |