Learn More
N-(4-Fluorophenyl)thiourea, 99%, Thermo Scientific™
Product Code. 11327596
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327596 1 g 1g
11337596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11327596 Supplier Thermo Scientific Alfa Aesar Supplier No. L11159.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 459-05-2
Molecular Formula C7H7FN2S
Molecular Weight (g/mol) 170.21
MDL Number MFCD00041180
InChI Key BRWKXKNZRVALNZ-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl thiourea, 1-4-fluorophenyl-2-thiourea, 4-fluorophenyl thiourea, n-4-fluorophenyl thiourea, p-fluorophenylthiourea, 4-fluoro-phenyl-thiourea, thiourea, 4-fluorophenyl, amino 4-fluorophenyl amino methane-1-thione, pubchem10059, acmc-1ajjb
PubChem CID 693061
IUPAC Name (4-fluorophenyl)thiourea
SMILES NC(=S)NC1=CC=C(F)C=C1
Specifications
Melting Point 162°C to 166°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 2803645
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material N-(4-Fluorophenyl)thiourea