Learn More
N-(4-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-(4-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine
Specifications
Specifications
| Chemical Name or Material | N-(4-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine |
| CAS | 1029721-32-1 |
| Analyte or Component Names | N-(4-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H23 N3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Quinazolinamine, N-(4-ethynylphenyl)-6,7-bis(2-methoxyethoxy)-, N-(4-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)-4-quinazolinamine, N-(4-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine |
| InChI Formula | InChI=1S/C22H23N3O4/c1-4-16-5-7-17(8-6-16)25-22-18-13-20(28-11-9-26-2)21(29-12-10-27-3)14-19(18)23-15-24-22/h1,5-8,13-15H,9-12H2,2-3H3,(H,23,24,25) |
| SMILES | COCCOc1cc2ncnc(Nc3ccc(cc3)C#C)c2cc1OCCOC |
| IUPAC Name | N-(4-ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine |
| Show More |