Learn More
N,4-Dimethylbenzenesulfonamide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 640-61-9 |
| Molecular Formula | C8H11NO2S |
| MDL Number | MFCD00008285 |
| Quantity | 1 kg |
| SMILES | O=S(C1=CC=C(C)C=C1)(NC)=O |
| Molecular Weight (g/mol) | 185.24 |
| Percent Purity | 97% |
| Chemical Name or Material | N,4-Dimethylbenzenesulfonamide |