Learn More
N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, TRC
Product Code. 30180360
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180360 1 g 1g
30179748 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180360 Supplier Toronto Research Chemicals Supplier No. TRCC3801301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2457316-06-0
Molecular Formula C16 H15 Cl N2 O3 S
Molecular Weight (g/mol) 350.82
Synonym N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, Benzenesulfonamide, 2-chloro-5-[(2,3-dihydro-2-methyl-1H-indol-1-yl)carbonyl]- (ACI), 2-Chloro-5-[(2,3-dihydro-2-methyl-1H-indol-1-yl)carbonyl]benzenesulfonamide (ACI)
IUPAC Name 2-chloro-5-(2-methyl-2,3-dihydroindole-1-carbonyl)benzenesulfonamide
SMILES CC1Cc2ccccc2N1C(=O)c3ccc(Cl)c(c3)S(=O)(=O)N
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H15ClN2O3S/c1-10-8-11-4-2-3-5-14(11)19(10)16(20)12-6-7-13(17)15(9-12)23(18,21)22/h2-7,9-10H,8H2,1H3,(H2,18,21,22)
Formula Weight 350.0492
Chemical Name or Material N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline