Learn More
N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, Mikromol™
Product Code. 30461329
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461329 4 x 25 mg 100mg
30049531 25 mg 25mg
30050390 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30461329 Supplier Mikromol Supplier No. MM0477.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline
CAS 2457316-06-0
Analyte or Component Names N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline
Shipping Condition Room Temperature
Molecular Formula C16 H15 Cl N2 O3 S
Certifications/Compliance ISO 17025
Synonym N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline
InChI Formula InChI=1S/C16H15ClN2O3S/c1-10-8-11-4-2-3-5-14(11)19(10)16(20)12-6-7-13(17)15(9-12)23(18,21)22/h2-7,9-10H,8H2,1H3,(H2,18,21,22)
Solution Type Neat
SMILES CC1Cc2ccccc2N1C(=O)c3ccc(Cl)c(c3)S(=O)(=O)N
IUPAC Name 2-chloro-5-(2-methyl-2,3-dihydroindole-1-carbonyl)benzenesulfonamide
Molecular Weight (g/mol) 350.82
Quantity 4 x 25 mg
Formula Weight 350.0492 g/mol