Learn More
N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline, Mikromol™
Description
N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline
Specifications
Specifications
| Chemical Name or Material | N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline |
| CAS | 2457316-06-0 |
| Analyte or Component Names | N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 Cl N2 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-(4-Chloro-3-sulfamoylbenzoyl)-2-methylindoline |
| InChI Formula | InChI=1S/C16H15ClN2O3S/c1-10-8-11-4-2-3-5-14(11)19(10)16(20)12-6-7-13(17)15(9-12)23(18,21)22/h2-7,9-10H,8H2,1H3,(H2,18,21,22) |
| SMILES | CC1Cc2ccccc2N1C(=O)c3ccc(Cl)c(c3)S(=O)(=O)N |
| IUPAC Name | 2-chloro-5-(2-methyl-2,3-dihydroindole-1-carbonyl)benzenesulfonamide |
| Show More |