missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Product Code. p-7038116
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493036 250 mg 250mg
15483036 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493036 Supplier Thermo Scientific Alfa Aesar Supplier No. H57015.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562080-79-9
Molecular Formula C14H9BrF3NO
Molecular Weight (g/mol) 344.13
MDL Number MFCD03937564
InChI Key JDUHFWWSDVISOZ-UHFFFAOYSA-N
Synonym n-4-bromophenyl-3-trifluoromethyl benzamide, benzamide, n-4-bromophenyl-3-trifluoromethyl, n-4-bromo-phenyl-3-trifluoromethyl-benzamide, n-4-bromophenyl 3-trifluoromethyl phenyl carboxamide
PubChem CID 531810
IUPAC Name N-(4-bromophenyl)-3-(trifluoromethyl)benzamide
SMILES FC(F)(F)C1=CC=CC(=C1)C(=O)NC1=CC=C(Br)C=C1

Specifications

Quantity 250 mg
Beilstein 14213316
Formula Weight 344.13
Percent Purity 97%
Chemical Name or Material N-(4-Bromophenyl)-3-(trifluoromethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.