missing translation for 'onlineSavingsMsg'
Learn More

N-[(4-Aminophenyl)methyl]adenosine, MedChemExpress

Code produit p-200098948
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Conditionnement:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
16489476 50 mg 50mg
16472788 10 mg 10mg
16464236 100 mg 100mg
16405537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16489476 Fournisseur Medchem Express Code fournisseur HY100130
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress N-[(4-Aminophenyl)methyl]adenosine is a adenosine receptor inhibitor, with Ki of 29 nM for Rat ecto-5′-Nucleotidase.

N-[(4-Aminophenyl)methyl]adenosine is a adenosine receptor inhibitor, with Ki of 29 nM for Rat ecto-5′-Nucleotidase.

Spécification

Chemical Name or Material N-[(4-Aminophenyl)methyl]adenosine
CAS 95523-13-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H20N6O4
Quantity 50 mg
Solubility Information DMSO : 10 mg/mL (26.85 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCC4=CC=C(N)C=C4)O1)O)O
Molecular Weight (g/mol) 372.38
Formula Weight 372.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.