Learn More
N-[(4-Aminophenyl)methyl]adenosine, MedChemExpress
Product Code. 16489476
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489476 50 mg 50mg
16472788 10 mg 10mg
16464236 100 mg 100mg
16405537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16489476 Supplier Medchem Express Supplier No. HY100130
 Restricted Item
Estimated Shipment: 16-11-2026
MedChemExpress N-[(4-Aminophenyl)methyl]adenosine is a adenosine receptor inhibitor, with Ki of 29 nM for Rat ecto-5′-Nucleotidase.
N-[(4-Aminophenyl)methyl]adenosine is a adenosine receptor inhibitor, with Ki of 29 nM for Rat ecto-5′-Nucleotidase.
Specifications
Chemical Name or Material N-[(4-Aminophenyl)methyl]adenosine
CAS 95523-13-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H20N6O4
Quantity 50 mg
Solubility Information DMSO : 10 mg/mL (26.85 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCC4=CC=C(N)C=C4)O1)O)O
Molecular Weight (g/mol) 372.38
Formula Weight 372.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation