Learn More
N-[(2S)-3-Amino-2-hydroxypropyl]-N-[3-fluoro-4-(4-morpholinyl)phenyl] acetamide Hydrochloride, TRC
Product Code. 30138174
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138174 5 mg 5mg
30008380 50 mg 50mg
30006087 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138174 Supplier Toronto Research Chemicals Supplier No. TRCA6090755MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1391068-25-9
Molecular Formula C15 H22 F N3 O3 . Cl H
Molecular Weight (g/mol) 347.81
Synonym Acetamide, N-[(2S)-3-amino-2-hydroxypropyl]-N-[3-fluoro-4-(4-morpholinyl)phenyl]-, hydrochloride (1:1), N-[(2S)-3-Amino-2-hydroxypropyl]-N-(3-fluoro-4-morpholinophenyl)acetamide Hydrochloride
IUPAC Name N-[(2S)-3-amino-2-hydroxypropyl]-N-(3-fluoro-4-morpholin-4-ylphenyl)acetamide;hydrochloride
SMILES Cl.CC(=O)N(C[C@@H](O)CN)c1ccc(N2CCOCC2)c(F)c1
Specifications
Analyte or Component Names N-[(2S)-3-Amino-2-hydroxypropyl]-N-(3-fluoro-4-morpholinophenyl)acetamide Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C15H22FN3O3.ClH/c1-11(20)19(10-13(21)9-17)12-2-3-15(14(16)8-12)18-4-6-22-7-5-18;/h2-3,8,13,21H,4-7,9-10,17H2,1H3;1H/t13-;/m0./s1
Solution Type Neat
Industry Type Forensics
Quantity 5 mg
Formula Weight 347.1412
Percent Purity >95%
Chemical Name or Material N-[(2S)-3-Amino-2-hydroxypropyl]-N-(3-fluoro-4-morpholinophenyl)acetamide Hydrochloride
Grade HPLC