Learn More
N-(2-Phenylethyl)thiourea, 97%, Thermo Scientific™
Product Code. 11308485
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308485 1 g 1g
11318485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11308485 Supplier Thermo Scientific Alfa Aesar Supplier No. L10520.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6815-00-5
Molecular Formula C9H12N2S
Molecular Weight (g/mol) 180.269
MDL Number MFCD00041194
InChI Key OFVGPHQYOCKLLM-UHFFFAOYSA-N
Synonym phenethylthiourea, 2-phenylethyl thiourea, n-phenethylthiourea, n-2-phenylethyl thiourea, phenethyl-thiourea, 1-phenethylthiourea, n-2-phenylethyl thioformamide, 1-2-phenylethyl thiourea, amino 2-phenylethyl amino methane-1-thione, 3-phenethylthiourea
PubChem CID 735845
IUPAC Name 2-phenylethylthiourea
SMILES C1=CC=C(C=C1)CCNC(=S)N
Specifications
Melting Point 136°C to 140°C
Odor Odorless
Linear Formula C6H5(CH2)2NHCSNH2
Quantity 1 g
Beilstein 2092343
Formula Weight 180.27
Percent Purity 97%
Chemical Name or Material N-(2-Phenylethyl)thiourea