Learn More
N-(2-Aminoethyl)benzamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-(2-Aminoethyl)benzamide |
| CAS | 1009-17-2 |
| Molecular Formula | C9H12N2O |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C9H12N2O/c10-6-7-11-9(12)8-4-2-1-3-5-8/h1-5H,6-7,10H2,(H,11,12) |
| SMILES | NCCNC(=O)c1ccccc1 |
| IUPAC Name | N-(2-aminoethyl)benzamide |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.095 |