Learn More
N-(2,6-Dichlorophenyl)isatin, TRC
Specifications
Specifications
| CAS | 24542-74-3 |
| Molecular Formula | C14 H7 Cl2 N O2 |
| Quantity | 50 mg |
| Synonym | 1-(2,6-Dichlorophenyl)indolin-2,3-dione |
| InChI Formula | InChI=1S/C14H7Cl2NO2/c15-9-5-3-6-10(16)12(9)17-11-7-2-1-4-8(11)13(18)14(17)19/h1-7H |
| SMILES | Clc1cccc(Cl)c1N2C(=O)C(=O)c3ccccc23 |
| IUPAC Name | 1-(2,6-dichlorophenyl)indole-2,3-dione |
| Molecular Weight (g/mol) | 292.12 |
| Formula Weight | 290.9854 |
| Chemical Name or Material | 1-(2,6-Dichlorophenyl)indolin-2,3-dione |