Learn More
N’-(2,4-Dimethylphenyl)-N-methylformamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N'-(2,4-Dimethylphenyl)-N-methylformamide Hydrochloride |
| CAS | 51550-40-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H14 N2 . Cl H |
| Quantity | 50 mg |
| Synonym | Methanimidamide, N-(2,4-dimethylphenyl)-N'-methyl-, hydrochloride (1:1), Methanimidamide, N-(2,4-dimethylphenyl)-N'-methyl-, monohydrochloride (9CI), Danjiami, Monocarboxoldehyde, N-(2,4-Dimethylphenyl)-N'-methylmethanimidamide hydrochloride, N-Methyl-N'-(2,4-xylyl)formamidine hydrochloride, N'-(2,4-Xylyl)-N-methylformamidine hydrochloride, Semiamitraz chloride, U 40481A |
| InChI Formula | InChI=1S/C10H14N2.ClH/c1-8-4-5-10(9(2)6-8)12-7-11-3;/h4-7H,1-3H3,(H,11,12);1H |
| SMILES | Cl.CN\C=N\c1ccc(C)cc1C |
| IUPAC Name | N'-(2,4-dimethylphenyl)-N-methylmethanimidamide;hydrochloride |
| Molecular Weight (g/mol) | 198.69 |
| Show More |