Learn More
N-(2,4-Dimethylphenyl)-2-chloroacetamide, TRC
Product Code. 30379651
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379651 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30379651 Supplier Toronto Research Chemicals Supplier No. TRCD476725500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(2,4-Dimethylphenyl)-2-chloroacetamide
CAS 39106-10-0
Molecular Formula C10H12ClNO
Quantity 500 mg
Synonym 2-Chloro-N-(2,4-dimethylphenyl)acetamide
InChI Formula InChI=1S/C10H12ClNO/c1-7-3-4-9(8(2)5-7)12-10(13)6-11/h3-5H,6H2,1-2H3,(H,12,13)
SMILES Cc1ccc(NC(=O)CCl)c(C)c1
Molecular Weight (g/mol) 197.66
Formula Weight 197.0607
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.