missing translation for 'onlineSavingsMsg'
Learn More

N{2-[4-(7-chloroquinolin-4-yl)piperzine-1-yl]-4,5-dihydroxy-6-(hydroxmethyl)oxan-3-yl}acetamide, TRC

Product Code. 30350898
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350898 50 mg 50mg
30354666 10 mg 10mg
30356245 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350898 Supplier Toronto Research Chemicals Supplier No. TRCC41562550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21 H27 Cl N4 O5
Molecular Weight (g/mol) 450.916
Synonym 7-Chloro-4-(piperazin-1-yl)quinoline N-Acetyl-b-D-glucosamine Adduct
IUPAC Name N-[2-[4-(7-chloroquinolin-4-yl)piperazin-1-yl]-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(O)C(O)C(CO)OC1N2CCN(CC2)c3ccnc4cc(Cl)ccc34

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H27ClN4O5/c1-12(28)24-18-20(30)19(29)17(11-27)31-21(18)26-8-6-25(7-9-26)16-4-5-23-15-10-13(22)2-3-14(15)16/h2-5,10,17-21,27,29-30H,6-9,11H2,1H3,(H,24,28)
Formula Weight 450.167
Chemical Name or Material N(2-[4-(7-chloroquinolin-4-yl)piperzine-1-yl]-4,5-dihydroxy-6-(hydroxmethyl)oxan-3-yl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.