Learn More
N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid tert-Butyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid tert-Butyl Ester |
| CAS | 1009093-14-4 |
| Molecular Formula | C14H21NO4 |
| Quantity | 1 g |
| Synonym | N-[(1R,2R)-2-Hydroxy-1-(hydroxymethyl)-2-phenylethyl]carbamic Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C14H21NO4/c1-14(2,3)19-13(18)15-11(9-16)12(17)10-7-5-4-6-8-10/h4-8,11-12,16-17H,9H2,1-3H3,(H,15,18)/t11-,12-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@H](CO)[C@H](O)c1ccccc1 |
| IUPAC Name | tert-butyl N-[(1R,2R)-1,3-dihydroxy-1-phenylpropan-2-yl]carbamate |
| Molecular Weight (g/mol) | 267.32 |
| Formula Weight | 267.1471 |