missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine, dihydrochoride, For ACS analysis, ≥98%, MP Biomedicals™

Product Code. 11472882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472882 10 g 10g
11482882 25 g 25g
11492882 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472882 Supplier MP Biomedicals Supplier No. 0215254910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Assay Percent Range ≥98%
Quantity 10 g
Formula Weight 259.18
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material N-(1-Naphthyl)Ethylenediamine), For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.