missing translation for 'onlineSavingsMsg'
Learn More

Myristoleic Acid Methyl Ester, 99%, MP Biomedicals™

Product Code. 11474951 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474951 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474951 Supplier MP Biomedicals Supplier No. 0215576180
  • ONLINE EXCLUSIVE PRICE
    112.00 EUR
    106.05 EUR / 100mg
    Save 5.95 EUR
    5% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56219-06-8
Assay Percent Range 99%
Molecular Formula C15H28O2
Quantity 100 mg
Synonym methyl myristoleate, myristoleic acid methyl ester, methyl cis-9-tetradecenoate, unii-m2a58gy964, z-methyl tetradec-9-enoate, methyl z-tetradec-9-enoate, cis-9-tetradecenoic acid, methyl ester, methyl 9z-tetradec-9-enoate, formyl 9z-tetradecenoate, methyl 9z-tetradecenoate
InChI Key RWIPSJUSVXDVPB-SREVYHEPSA-N
SMILES CCCCC=CCCCCCCCC(=O)OC
IUPAC Name methyl (Z)-tetradec-9-enoate
Molecular Weight (g/mol) 240.387
PubChem CID 5352674
Formula Weight 240.4
Physical Form Liquid
Chemical Name or Material Myristoleic acid methyl ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.